Learn More
Cell Biolabs Inc ViraSafe™ Pantropic Lentiviral Packaging System
Catalog No. NC9056248
Encompass_Preferred
Cell Biolabs' ViraSafe™ Lentiviral Expression and Packaging Systems are third-generation lentiviral systems designed to package recombinant lentivirus with additional built-in safety features to further minimize the chance of producing replication-competent virus. The ViraSafe™ Pantropic Lentiviral Packaging System contains three packaging plasmids for co-transfection with any third generation lentiviral expression vector of your choice. Pantropic lentiviruses made from this system contain VSVG envelope and readily infect cells of any species.
Catalog No. NC9056248 Supplier Cell Biolabs Inc Supplier No. VPK206
Cell Biolabs' ViraSafe™ Lentiviral Expression and Packaging Systems are third-generation lentiviral systems designed to package recombinant lentivirus with additional built-in safety features to further minimize the chance of producing replication-competent virus. The ViraSafe™ Pantropic Lentiviral Packaging System contains three packaging plasmids for co-transfection with any third generation lentiviral expression vector of your choice. Pantropic lentiviruses made from this system contain VSVG envelope and readily infect cells of any species.