Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Virchow Brain Sectioning Knife, Double-Edged

Catalog No. 21124624 Shop All Surgical Design Products
Change view
Click to view available options
Size:
180 x 20 mm
250 x 40 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-624 250 x 40 mm
21-124-623 180 x 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-624 Supplier Surgical Design™ Supplier No. ATPM004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Virchow brain sectioning knife delivers precise, uniform slices for neuropathological and postmortem brain examination.

  • Double-edged stainless steel blade provides exceptionally smooth and uniform cuts through brain tissue
  • Available in 80 x 20 mm and 250 x 40 mm blade sizes to support both fine and large-format sectioning
  • Designed specifically for neuropathology, forensic pathology, and anatomical lab dissection
  • Rigid, precision-ground blade maintains stability for consistent slice thickness
  • Ergonomic handle ensures safe, controlled movement during delicate cerebral examination
  • Ideal for coronal, sagittal, and axial brain sectioning during postmortem analysis
  • Highly polished stainless surface resists corrosion and cleans easily even after extended autopsy use
  • Balanced weight distribution reduces fatigue when processing multiple brain specimens
  • Autoclavable and reusable, compatible with hospital, forensic, and research laboratory sterilization protocols
  • Gross brain examination in forensic and hospital autopsies
  • Neuropathological evaluation for trauma, hemorrhage, degenerative disease, and neoplasms
  • Production of diagnostic slices for microscopy, photography, and case documentation

Specifications

Product Type Autopsy Dissecting Knife
Size 250 x 40 mm
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO/GMP
Autoclavable Autoclavable
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.