Learn More
World Precision Instrument Virchow Skull Breaker, 7.6cm
SDP
Catalog No. 502538384
Encompass
The Virchow Skull Breaker is a T-handled chisel. After using a bone saw to divide the cranial bones, you can use this blade to pry them apart during a post-mortem exam. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8384 Supplier World Precision Instrument Supplier No. WP0004
The Virchow Skull Breaker is a T-handled chisel. After using a bone saw to divide the cranial bones, you can use this blade to pry them apart during a post-mortem exam. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.