Learn More
MilliporeSigma Viresolve Pro Micro Shield H Kit
Catalog No. NC1515794
Encompass
The Viresolve Pro PD kit is ideally suited for performing your evaluation, optimization and scaling studies. Each kit contains 9 Micro devices from one membrane layering combination, a user guide and certificate of quality. The air vent prevents air locking within the device.
Catalog No. NC1515794 Supplier MilliporeSigma Supplier No. VPMHKITNB9
The Viresolve Pro PD kit is ideally suited for performing your evaluation, optimization and scaling studies. Each kit contains 9 Micro devices from one membrane layering combination, a user guide and certificate of quality. The air vent prevents air locking within the device.