Learn More
MilliporeSigma Viresolve Pro Micro Shield Kit
Catalog No. NC0473510
Encompass
The Viresolve Pro PD kit is ideally suited for performing your evaluation, optimization and scaling studies. Each kit contains 9 Micro devices from one membrane layering combination, a user guide and certificate of quality. The air vent prevents air locking within the device.
This item is not returnable. View return policy
Catalog No. NC0473510 Supplier MilliporeSigma Supplier No. VPMSKITNB9
The Viresolve Pro PD kit is ideally suited for performing your evaluation, optimization and scaling studies. Each kit contains 9 Micro devices from one membrane layering combination, a user guide and certificate of quality. The air vent prevents air locking within the device.