Learn More
MSC Diversey All-Purpose Disinfectant Cleaner, Lemon Scent, EPA Registered, 1 gal
Catalog No. 502868618
Encompass
Diversey All-Purpose Disinfectant Cleaner is a liquid solution designed for commercial and industrial use. It effectively cleans and disinfects various surfaces, targeting a broad spectrum of pathogens including viruses, fungi, tuberculosis, MRSA, and VRE.
  • Effective against a broad spectrum of bacteria, viruses, and fungi
  • Suitable for use on porcelain, plastic, floors, and walls
  • EPA registered disinfectant with a lemon/citrus scent
  • Clear, light liquid with a pH greater than 11
  • Supplied in a 1 gal bottle for versatile applications
Catalog No. 50-286-8618 Supplier MSC Supplier No. 10725984
Diversey All-Purpose Disinfectant Cleaner is a liquid solution designed for commercial and industrial use. It effectively cleans and disinfects various surfaces, targeting a broad spectrum of pathogens including viruses, fungi, tuberculosis, MRSA, and VRE.
  • Effective against a broad spectrum of bacteria, viruses, and fungi
  • Suitable for use on porcelain, plastic, floors, and walls
  • EPA registered disinfectant with a lemon/citrus scent
  • Clear, light liquid with a pH greater than 11
  • Supplied in a 1 gal bottle for versatile applications