Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Virex™ II 256 One-Step Disinfectant Cleaner and Deodorant

Catalog No. 17000040
Change view
Click to view available options
Volume (Metric):
946 mL
2.5 L
3.78 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17-000-040 946 mL
17-000-039 3.78 L
17-000-041 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-000-040 Supplier Diversey™ Supplier No. 04331.
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A one-step, quaternary disinfectant cleaner and deodorant to clean and disinfect hard surfaces.

  • Kills microorganisms including HIV-1, VRE, MRSA, GRSA, MRSE, VISA, PRSP, Herpes Simplex Types 1&2, Influenza Type A2, Adenovirus, Rotavirus and many more
  • Kills the organisms that cause odors and works as an odor counteractant to further eliminate odors not associated with bacteria
  • Highly concentrated quaternary formula provides excellent one-step, cost-effective cleaning and disinfection

Specifications

Product Type Quat-based Disinfectant
pH 10.2
Sterility Non-sterile
Appearance Blue
Certifications/Compliance EPA, FDA
Container Type AccuMix™ Container
Quantity 6 Pk.
Volume (English) 32 oz.
Volume (Metric) 946 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.