Learn More
Medchemexpress LLC HY-N6686 5mg Medchemexpress, Virginiamycin M1 CAS:21411-53-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66865MG
Encompass_Preferred
Medchemexpress, HY-N6686 5mg Virginiamycin M1 CAS:21411-53-0 Pristinamycin IIA (RP 12536) is a macrocyclic lactone peptolide antibiotic, derived from Streptomyces pristinaespiralis, which is a member of the streptogramin A group of antibiotics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2956