Learn More
Enzo Life Sciences Virginiamycin M1 (5mg). CAS: 21411-53-0
SDP
Catalog No. 502013379
Encompass_Preferred
Antibiotic that inhibits bacterial protein synthesis. Alternative name: Mikamycin A, Staphilomycin, Ostreogrycin A. Purity: ≥95% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-201-3379 Supplier Enzo Life Sciences Supplier No. ENZCHM1050005
Antibiotic that inhibits bacterial protein synthesis. Alternative name: Mikamycin A, Staphilomycin, Ostreogrycin A. Purity: ≥95% (HPLC). Long Term Storage: -20°C.