Learn More
Enzo Life Sciences Virginiamycin S1 (5mg). CAS: 23152-29-6
SDP
Supplier:  Enzo Life Sciences ENZCHM1060005
Encompass_Preferred
Antibiotic that inhibits bacterial protein synthesis. Alternative name: Staphilomycin S. Purity: ≥99% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-201-3380