Learn More
AdipoGen Viridicatumtoxin
SDP
Supplier:  AdipoGen AGCN20172M001
Encompass_Preferred
Chemical. CAS 39277-41-3. Formula C30H31NO10. MW 565.6. Isolated from Penicillium brasilianum. Mycotoxin. Tetracycline antibiotic. Potent antibacterial agent with activity against several strains of Staphylococcus aureus, including Methicillin-resistant S. aureus MRSA and quinolone-resistant S. aureus QRSA. Inhibits bacterial undecaprenyl pyrophosphate UPP synthase activity. Modest antitumor agent against selected cancer cell lines IC50~1µM.
Catalog No. 50-256-2645