Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viridiflorol, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014589 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4589 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4589 Supplier MilliporeSigma Supelco Supplier No. 7299910MG
Add to Cart
Add to Cart

Viridiflorol belongs to the class of aromadendrene sesquiterpenoids, isolated from the leaves of Salvia officinalis and detected in leaves of Salvia triloba.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 552-02-3
Linear Formula C15H26 O
MDL Number MFCD28899097
Synonym (1 aR,4 S,4 AS,7 R,7 AS)-1,1,4,7-Tetramethyl-2,3,4 A,5,6,7,7 A,7 b-octahydro-1 aH-cyclopropa[e]azulen-4-ol; [1 aR-(1 alpha,4 beta,4 beta,7 alpha,7 abeta,7 balpha)]-Decahydro-1,1,4,7-tetramethyl-1 H-cycloprop[e]azulen-4-ol
Type Analytical Standard
Quantity 10 mg
Formula Weight 222.37
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.