Learn More
AdipoGen Viridiol (250 µg)
SDP
Supplier:  AdipoGen AGCN20126C250
Encompass_Preferred
Chemical. CAS: 23820-80-6. Formula: C20H18O6. MW: 354.4. Isolated from Trichoderma sp. Steroidal antibiotic. Belongs to the Viridin family, including Wortmannin (Prod. No. AG-CN2-0023 http://www.adipogen.com/ag-cn2-0023/wortmannin.html ). Antifungal. Phytotoxin. Shows necrotic activity on plants. Phosphatidylinositol 3-kinase (PI3K) inhibitor.
Catalog No. 50-205-3497