Learn More
Waters Corp Waters Viridis BEH 2-Ethylpyridine Column, 130Å, 5 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp 186005757
Encompass_Preferred
The Viridis BEH 2-Ethylpyridine Column is engineered for SFC and HPLC, providing excellent selectivity and efficiency for diverse compound separation. It’s ideal for pharmaceutical, biomedical, environmental, and chemical analysis, offering exceptional stability and reproducible results with various mobile phases.
  • Designed for SFC; wide selectivity.
  • Excellent peak shapes without additives.
  • 130Å pore, 5 μm particle size.
  • Ample sample capacity; excellent resolution.
  • Exceptional stability and reproducibility.
  • Innovative bonded phase enhances retention.
  • Supports reverse-phase and normal-phase modes.
  • Compatible with aqueous and organic solvents.
  • High pressure tolerance; long column life.
Catalog No. 50-466-241