Learn More
Waters Corp Viridis BEH 2-Ethylpyridine Column, 130Å, 5 μm, 4.6 x 100 mm
Catalog No. 50466239 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis BEH 2-Ethylpyridine Column is engineered for chromatographic applications, providing good retention, improved peak shape, and unique selectivity.<ul><li>Good retention, improved peak shape, and unique selectivity</li><li>Accommodates increased mass loads</li><li>Scalable and available in analytical and preparative column dimensions</li><li>Internal diameter: 4.6 mm</li><li>Length: 100 mm</li><li>Particle size: 5 μm</li><li>Phase: Viridis BEH 2-Ethylpyridine</li><li>Pore size: 130 Å</li></ul>
Catalog No. 50-466-239 Supplier Waters Corp Supplier No. 186005755
The Viridis BEH 2-Ethylpyridine Column is engineered for chromatographic applications, providing good retention, improved peak shape, and unique selectivity.<ul><li>Good retention, improved peak shape, and unique selectivity</li><li>Accommodates increased mass loads</li><li>Scalable and available in analytical and preparative column dimensions</li><li>Internal diameter: 4.6 mm</li><li>Length: 100 mm</li><li>Particle size: 5 μm</li><li>Phase: Viridis BEH 2-Ethylpyridine</li><li>Pore size: 130 Å</li></ul>