Learn More
Waters Corp Waters Column, 2-Ethylpyridine, 5 μm, 3 x 150 mm, 130 Å
Catalog No. 50466236 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-performance column is designed for Supercritical Fluid Chromatography (SFC) applications, featuring a 3.0 x 150 mm dimension and utilizing 5 μm particle size BEH 2-Ethylpyridine chemistry. It demonstrates a wide range of selectivity for greater resolution, delivering excellent peak shapes under SFC conditions without the need for mobile-phase additives, even for well-retained basic achiral compounds.
  • Designed and tested for SFC applications.
  • Demonstrates a wide range of selectivity for greater resolution.
  • Features reduction and control of surface silanol activity.
  • Delivers excellent peak shapes under SFC conditions.
  • Eliminates the need for mobile-phase additives.
  • Suitable for well-retained basic achiral compounds.
  • Utilizes BEH (Ethylene Bridged Hybrid) particle technology.
Catalog No. 50-466-236 Supplier Waters Corp Supplier No. 186005752
This high-performance column is designed for Supercritical Fluid Chromatography (SFC) applications, featuring a 3.0 x 150 mm dimension and utilizing 5 μm particle size BEH 2-Ethylpyridine chemistry. It demonstrates a wide range of selectivity for greater resolution, delivering excellent peak shapes under SFC conditions without the need for mobile-phase additives, even for well-retained basic achiral compounds.
  • Designed and tested for SFC applications.
  • Demonstrates a wide range of selectivity for greater resolution.
  • Features reduction and control of surface silanol activity.
  • Delivers excellent peak shapes under SFC conditions.
  • Eliminates the need for mobile-phase additives.
  • Suitable for well-retained basic achiral compounds.
  • Utilizes BEH (Ethylene Bridged Hybrid) particle technology.