Learn More
Waters Corp OBD Prep Column, 2-Ethylpyridine, 5 μm, 130 Å, 10 mm x 100 mm
Catalog No. 502390521 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis BEH 2-Ethylpyridine OBD Prep Column combines advanced media manufacturing with industry-leading technology, offering high reproducibility for laboratory-scale purification. It utilizes patented Optimum Bed Density (OBD) Technology to enhance mechanical stability and extend column lifetimes, suitable for both analytical and preparative supercritical fluid chromatography (SFC).
  • Provides enhanced reproducibility for laboratory-scale purification.
  • Utilizes patented OBD technology for enhanced mechanical stability and extended column lifetimes.
  • Designed for supercritical fluid chromatography (SFC) in both analytical and preparative applications.
  • Features 2-Ethylpyridine chemistry with 5 μm spherical particles and 130 Å pore size.
  • Column dimensions: 10 mm inner diameter and 100 mm length.
  • Withstands maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-239-0521 Supplier Waters Corp Supplier No. 186008257
The Viridis BEH 2-Ethylpyridine OBD Prep Column combines advanced media manufacturing with industry-leading technology, offering high reproducibility for laboratory-scale purification. It utilizes patented Optimum Bed Density (OBD) Technology to enhance mechanical stability and extend column lifetimes, suitable for both analytical and preparative supercritical fluid chromatography (SFC).
  • Provides enhanced reproducibility for laboratory-scale purification.
  • Utilizes patented OBD technology for enhanced mechanical stability and extended column lifetimes.
  • Designed for supercritical fluid chromatography (SFC) in both analytical and preparative applications.
  • Features 2-Ethylpyridine chemistry with 5 μm spherical particles and 130 Å pore size.
  • Column dimensions: 10 mm inner diameter and 100 mm length.
  • Withstands maximum pressure of 6000 psi (415 Bar).