Learn More
Waters Corp Viridis OBD Prep Column, 2-Ethylpyridine, 130Å, 5 μm
Catalog No. 502391035 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Supercritical Fluid Chromatography (SFC) columns are engineered for enhanced reproducibility in laboratory-scale purification. They integrate state-of-the-art media manufacturing with industry-leading column technology, suitable for analytical and preparative chromatography.
  • Utilizes Optimum Bed Density (OBD) Technology for mechanical stability and extended column lifespan.
  • Offers achiral SFC selectivities with particle technologies like Ethylene Bridged Hybrid (BEH) and Charged Surface Hybrid (CSH).
  • Achieves excellent peak geometries for basic achiral chemicals under SFC conditions by reducing surface silanol activity.
  • Produced in specialized facilities adhering to cGMP and ISO 9001 standards, ensuring high-quality manufacturing.
Catalog No. 50-239-1035 Supplier Waters Corp Supplier No. 186008259
Add to Cart
Add to Cart
These Supercritical Fluid Chromatography (SFC) columns are engineered for enhanced reproducibility in laboratory-scale purification. They integrate state-of-the-art media manufacturing with industry-leading column technology, suitable for analytical and preparative chromatography.
  • Utilizes Optimum Bed Density (OBD) Technology for mechanical stability and extended column lifespan.
  • Offers achiral SFC selectivities with particle technologies like Ethylene Bridged Hybrid (BEH) and Charged Surface Hybrid (CSH).
  • Achieves excellent peak geometries for basic achiral chemicals under SFC conditions by reducing surface silanol activity.
  • Produced in specialized facilities adhering to cGMP and ISO 9001 standards, ensuring high-quality manufacturing.