Learn More
Waters Corp Waters Preparative Column, 2-Ethylpyridine, 5 μm, 19 mm, 100 mm
Supplier:  Waters Corp 186005763
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SFC
  • Offers a wide range of selectivity for greater resolution
  • Features reduction and control of surface silanol activity on Hybrid particles
  • Delivers excellent peak shapes under SFC conditions without the use of mobile-phase additives
  • Effective even for well-retained basic achiral compounds
Catalog No. 50-466-247