Learn More
Waters Corp Waters Preparative SFC Column, OBD 19 x 50 mm, 5 μm, BEH 2-EP
Catalog No. 50466246 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for preparative Supercritical Fluid Chromatography (SFC) applications. It is suitable for preparatory scale separations and is generally used for purifying larger quantities of target compounds.
  • Preparative SFC column
  • BEH 2-EP (Ethylene Bridged Hybrid 2-Ethylpyridine) chemistry
  • 5 μm particle size
  • OBD (Optimum Bed Density) 19 x 50 mm dimensions
Catalog No. 50-466-246 Supplier Waters Corp Supplier No. 186005762
This column is designed for preparative Supercritical Fluid Chromatography (SFC) applications. It is suitable for preparatory scale separations and is generally used for purifying larger quantities of target compounds.
  • Preparative SFC column
  • BEH 2-EP (Ethylene Bridged Hybrid 2-Ethylpyridine) chemistry
  • 5 μm particle size
  • OBD (Optimum Bed Density) 19 x 50 mm dimensions