Learn More
Waters Corp Viridis BEH 2-Ethylpyridine OBD Prep Column, 130Å, 5 µm, 50 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186005773
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 µm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-257
Add to Cart
Request a Quote