Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Viridis BEH Prep Guard Cartridge, 2-Ethylpyridine, 5 μm, 130 Å

Supplier:  Waters Corp 186006909

Encompass_Preferred

The 30x10mm Prep Guard Cartridge is designed to safeguard both HPLC and SFC 30 mm ID Prep OBD™ columns. Installing this guard assembly before the prep column extends column lifetimes and reduces purification costs.

  • Features 2-Ethylpyridine chemistry
  • Has a particle size of 5 μm
  • Pore size of 130 Å
  • Designed for SuperCritical Fluid (SFC) separation mode
  • Compatible with SFC and SFC/MS techniques
  • Utilizes BEH particle technology
  • Withstands maximum pressure of 6000 psi (415 Bar)
  • Has an inner diameter of 30 mm and length of 10 mm

Catalog No. 50-239-2344


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.