Learn More
Waters Corp Waters Viridis BEH Column, 130 Å, 5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186006544
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SFC.
  • Demonstrate a wide range of selectivity for greater resolution.
  • Reduction and control of surface silanol activity on hybrid particles.
  • Delivers excellent peak shapes under SFC conditions, without the use of mobile-phase additives, even for well-retained basic achiral compounds.
Catalog No. 50-466-307