Learn More
Waters Corp Waters Viridis BEH Column, 5 μm, 130 Å, 4.6 × 100 mm
Supplier:  Waters Corp 186005724
Encompass_Preferred
The Viridis BEH Column is an analytical SFC column based on patented Ethylene Bridged Hybrid (BEH) particle technology. It is widely used in achiral SFC separations, providing good retention, peak shape, and selectivity.
  • Spherical particle shape
  • 0.7 cc/g pore volume
  • 185 m2/g surface area
  • Unbonded chemistry
  • Provides good retention, peak shape, and selectivity both with and without the use of additives
Catalog No. 50-466-214