Learn More
Waters Corp Viridis OBD Prep Column, 130Å, 5 μm, 30 mm, 250 mm
Catalog No. 50466229 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SuperCritical Fluid (SFC) applications.
  • Offers a wide range of selectivity for enhanced resolution.
  • Features reduction and control of surface silanol activity on hybrid particles.
  • Delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without requiring mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.
Catalog No. 50-466-229 Supplier Waters Corp Supplier No. 186005739
Add to Cart
Add to Cart
Request a Quote
Designed and tested for SFC, the Viridis 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SuperCritical Fluid (SFC) applications.
  • Offers a wide range of selectivity for enhanced resolution.
  • Features reduction and control of surface silanol activity on hybrid particles.
  • Delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without requiring mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.