Learn More
Waters Corp Prep Column, Viridis BEH, 5 μm, 30 x 150 mm
Catalog No. 50466228 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Viridis columns provide an extended array of achiral SFC selectivities. They are built upon the patented Ethylene Bridged Hybrid (BEH) particle technology. Through the reduction and regulation of surface silanol activity on Viridis particles, these columns achieve excellent peak shapes, even for well-retained basic achiral compounds under SFC conditions. Separation methods can be optimized and scaled up to Viridis Prep OBD™ Columns.
  • Offers an added range of achiral SFC selectivities.
  • Based on patented Ethylene Bridged Hybrid (BEH) particle technology.
  • Reduction and control of surface silanol activity on Viridis particles.
  • Delivers excellent peak shapes for well-retained basic achiral compounds under SFC conditions.
  • Designed for optimizing and scaling up separation methods.
Catalog No. 50-466-228 Supplier Waters Corp Supplier No. 186005738
Add to Cart
Add to Cart
Request a Quote
Viridis columns provide an extended array of achiral SFC selectivities. They are built upon the patented Ethylene Bridged Hybrid (BEH) particle technology. Through the reduction and regulation of surface silanol activity on Viridis particles, these columns achieve excellent peak shapes, even for well-retained basic achiral compounds under SFC conditions. Separation methods can be optimized and scaled up to Viridis Prep OBD™ Columns.
  • Offers an added range of achiral SFC selectivities.
  • Based on patented Ethylene Bridged Hybrid (BEH) particle technology.
  • Reduction and control of surface silanol activity on Viridis particles.
  • Delivers excellent peak shapes for well-retained basic achiral compounds under SFC conditions.
  • Designed for optimizing and scaling up separation methods.