Learn More
Waters Corp Viridis BEH OBD Prep Column, 130Å, 5 μm, 19 mm x 250 mm
Catalog No. 50466224 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column is designed for SuperCritical Fluid Chromatography (SFC), offering broad selectivity and enhanced resolution. It incorporates Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies for consistent performance. Its optimized design and manufacturing ensure increased lifetime and reproducibility, addressing mechanical instabilities.
  • Designed for SuperCritical Fluid (SFC) applications.
  • Provides broad selectivity and enhanced resolution.
  • Delivers excellent peak shapes without mobile-phase additives under SFC conditions.
  • Features Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies.
  • Ensures batch-to-batch reproducibility.
  • Optimized packing and unique design increase lifetime and reproducibility.
  • Manufactured with high purity materials for consistent performance.
Catalog No. 50-466-224 Supplier Waters Corp Supplier No. 186005734
Add to Cart
Add to Cart
Request a Quote
This preparative column is designed for SuperCritical Fluid Chromatography (SFC), offering broad selectivity and enhanced resolution. It incorporates Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies for consistent performance. Its optimized design and manufacturing ensure increased lifetime and reproducibility, addressing mechanical instabilities.
  • Designed for SuperCritical Fluid (SFC) applications.
  • Provides broad selectivity and enhanced resolution.
  • Delivers excellent peak shapes without mobile-phase additives under SFC conditions.
  • Features Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies.
  • Ensures batch-to-batch reproducibility.
  • Optimized packing and unique design increase lifetime and reproducibility.
  • Manufactured with high purity materials for consistent performance.