Learn More
Waters Corp Waters Viridis BEH OBD Prep Column, 130Å, 5 μm, 50 mm × 150 mm
Supplier:  Waters Corp 186005742
Encompass_Preferred
Designed and tested for SuperCritical Fluid (SFC), the Viridis 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SuperCritical Fluid conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SuperCritical Fluid (SFC)
  • Demonstrate a wide range of selectivity for greater resolution
  • Reduction and control of surface silanol activity on Viridis Hybrid particles delivers excellent peak shapes under SuperCritical Fluid conditions
  • Excellent peak shapes without the use of mobile-phase additives, even for well-retained basic achiral compounds
Catalog No. 50-466-232
Add to Cart
Request a Quote