Learn More
Waters Corp Viridis BEH OBD Prep Column, 130Å, 5μm, 50 x 250 mm
Supplier:  Waters Corp 186005743
Encompass_Preferred
This Viridis BEH OBD Prep Column is designed to optimize preparative liquid chromatography (Prep LC) applications. It features a robust hybrid silica phase with a 130 Å pore size and 5 μm particle size, ensuring direct scalability from analytical to preparative methods and delivering consistent, high-quality results.
  • Bridges the performance gap in Prep LC
  • Uniform, optimum packed bed density
  • Direct scalability from analytical to preparative methods
  • Excellent column lifetime and consistent performance
  • Good resolution, peak shape, and mass loading
  • Universal choice for various compounds
  • Exceptional peak shape for basic analytes at elevated pH
  • Stable across wide pH range and high temperatures
  • Maximizes recoveries with robust packings
  • Decreases risk of additional Prep method development
Catalog No. 50-466-233
Add to Cart
Request a Quote