Learn More
Waters Corp Waters CSH Fluoro-Phenyl Column, 130Å, 5 μm, 3 mm X 150 mm
Catalog No. 50466186 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Viridis CSH Fluoro-Phenyl columns offer an added range of achiral SFC selectivities. These columns are based on Charged Surface Hybrid (CSH™) particle technology. The reduction and control of surface silanol activity on particles ensures excellent peak shapes even for well-retained basic achiral compounds under SFC conditions. The columns provide a wide range of selectivities based on particle and ligand technologies.
  • Based on charged surface hybrid (CSH™) particle technology
  • Offers an added range of achiral SFC selectivities
  • Reduction and control of surface silanol activity on particles results in excellent peak shapes
  • Suitable for well-retained basic achiral compounds under SFC conditions
  • Provides a wide range of selectivities based on particle and ligand technologies
Catalog No. 50-466-186 Supplier Waters Corp Supplier No. 186005690
Viridis CSH Fluoro-Phenyl columns offer an added range of achiral SFC selectivities. These columns are based on Charged Surface Hybrid (CSH™) particle technology. The reduction and control of surface silanol activity on particles ensures excellent peak shapes even for well-retained basic achiral compounds under SFC conditions. The columns provide a wide range of selectivities based on particle and ligand technologies.
  • Based on charged surface hybrid (CSH™) particle technology
  • Offers an added range of achiral SFC selectivities
  • Reduction and control of surface silanol activity on particles results in excellent peak shapes
  • Suitable for well-retained basic achiral compounds under SFC conditions
  • Provides a wide range of selectivities based on particle and ligand technologies