Learn More
Waters Corp Waters Viridis CSH Fluoro-Phenyl Column, 5 μm, 3.0 x 250 mm
Catalog No. 50466187 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis CSH Fluoro-Phenyl Column is designed for Supercritical Fluid Chromatography (SFC) applications. It utilizes CSH™ particle technology and offers excellent peak shapes, particularly for well-retained basic achiral compounds.
  • Uses CSH™ particle technology for SFC applications.
  • Provides excellent peak shapes for basic achiral compounds.
  • Features a particle size of 5 μm.
  • Has dimensions of 3.0 x 250 mm.
  • Based on patented Charged Surface Hybrid particle technology derived from Ethylene Bridged Hybrid particle technology.
Catalog No. 50-466-187 Supplier Waters Corp Supplier No. 186005691
Add to Cart
Add to Cart
The Viridis CSH Fluoro-Phenyl Column is designed for Supercritical Fluid Chromatography (SFC) applications. It utilizes CSH™ particle technology and offers excellent peak shapes, particularly for well-retained basic achiral compounds.
  • Uses CSH™ particle technology for SFC applications.
  • Provides excellent peak shapes for basic achiral compounds.
  • Features a particle size of 5 μm.
  • Has dimensions of 3.0 x 250 mm.
  • Based on patented Charged Surface Hybrid particle technology derived from Ethylene Bridged Hybrid particle technology.