Learn More
Waters Corp Waters Column, PFP/Fluoro-Phenyl, 5 μm, 130 Å, 4.6 mm x 150 mm
Supplier:  Waters Corp 186005694
Encompass_Preferred
Designed and tested for SFC, these 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for SFC.
  • Demonstrates a wide range of selectivity for greater resolution.
  • Reduction and control of surface silanol activity on Hybrid particles.
  • Delivers excellent peak shapes under SFC conditions without mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.
Catalog No. 50-466-190