Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SFC Column, 130Å, 5 μm, 10 × 100 mm

Supplier:  Waters Corp 186008249

Encompass_Preferred

This SFC column is designed for preparative supercritical fluid chromatography (SFC) applications. It utilizes advanced Viridis CSH Fluoro-Phenyl OBD Prep technology, featuring specific pore and particle sizes, and precise dimensions for laboratory-scale separations.

  • Utilizes Viridis CSH Fluoro-Phenyl OBD Prep technology.
  • Features a 130Å pore size.
  • Designed with a 5 μm particle size.
  • Dimensions are 10 × 100 mm.
  • Suitable for preparative SFC applications.

Catalog No. 50-239-1606


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.