Learn More
Waters Corp Waters Corporation Prep Column, CSH, Fluoro-Phenyl, 5 μm, 30 mm × 100 mm
Supplier:  Waters Corp 186005706
Encompass_Preferred
Designed and tested for SFC, these 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • PFP/Fluoro-Phenyl packing and bonding material
  • Hybrid particle matrix with CSH technology
  • Supercritical fluid (SFC) separation mode and SFC/MS technique compatibility
  • 5 μm particle size and 130 Å pore size
  • 30 mm inner diameter and 100 mm length
  • Maximum pressure of 6000 psi (415 Bar)
  • Medium silanol activity and spherical particle shape
  • 10% carbon load
  • Not endcapped
  • One unit per package
Catalog No. 50-466-202
Add to Cart
Request a Quote