Learn More
Waters Corp Viridis CSH Fluoro-Phenyl OBD Prep Column, 130Å, 5 µm, 19 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186005702
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 µm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-198