Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SFC Column, Viridis Silica 2-EP, 5 μm, 4.6 x 150 mm

Supplier:  Waters Corp 186004937

Encompass_Preferred

This SFC column is designed for achiral separations, offering excellent retention, peak shape, and selectivity.

  • Dimensions: 4.6 x 150 mm
  • Particle size: 5 μm
  • Optimized for achiral SFC separations
  • Delivers good retention, peak shape, and selectivity

Catalog No. 50-791-246


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.