Learn More
Waters Corp Waters Prep Column, 2-Ethylpyridine, 5 μm, 100 Å, 30 mm × 50 mm
Catalog No. 50791254 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Combining state-of-the-art media manufacturing with industry-leading column technology, Supercritical Fluid Chromatography (SFC) columns offer a new level of reproducibility for laboratory-scale purification. SFC columns are available for analytical and preparative chromatography as SFC 2-Ethylpyridine columns and SFC Silica columns. Larger preparative column formats are manufactured using patented Optimum Bed Density (OBD) Technology.
  • The 2-Ethylpyridine chemistry is successfully used for a wide range of achiral SFC applications due to its retention, peak shape, and selectivity properties.
  • Columns also provide high reliability and reproducibility and can be used for analytical and preparative applications.
  • Based on long history of chromatographic silica production, the silica columns are designed to be highly reproducible and predictable, based on tight product specifications and very low metal content.
Catalog No. 50-791-254 Supplier Waters Corp Supplier No. 186004947
Combining state-of-the-art media manufacturing with industry-leading column technology, Supercritical Fluid Chromatography (SFC) columns offer a new level of reproducibility for laboratory-scale purification. SFC columns are available for analytical and preparative chromatography as SFC 2-Ethylpyridine columns and SFC Silica columns. Larger preparative column formats are manufactured using patented Optimum Bed Density (OBD) Technology.
  • The 2-Ethylpyridine chemistry is successfully used for a wide range of achiral SFC applications due to its retention, peak shape, and selectivity properties.
  • Columns also provide high reliability and reproducibility and can be used for analytical and preparative applications.
  • Based on long history of chromatographic silica production, the silica columns are designed to be highly reproducible and predictable, based on tight product specifications and very low metal content.