Learn More
Waters Corp WATERS CORPORATION Viridis SFC Column, 2-Ethylpyridine OBD, 30 x 75 mm, 5 μm
Catalog No. 50791255 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Viridis SFC 2-Ethylpyridine OBD Columns are designed for a wide range of achiral SFC applications, offering excellent peak shape, high mass loading, reliability, and reproducibility for both analytical and preparative uses.
  • Excellent peak shape
  • High mass loading capabilities
  • Consistent reliability and reproducibility
  • Suitable for both analytical and preparative applications
  • Features 2-ethylpyridine ligand chemistry
Catalog No. 50-791-255 Supplier Waters Corp Supplier No. 186004948
Add to Cart
Add to Cart
Request a Quote
Viridis SFC 2-Ethylpyridine OBD Columns are designed for a wide range of achiral SFC applications, offering excellent peak shape, high mass loading, reliability, and reproducibility for both analytical and preparative uses.
  • Excellent peak shape
  • High mass loading capabilities
  • Consistent reliability and reproducibility
  • Suitable for both analytical and preparative applications
  • Features 2-ethylpyridine ligand chemistry