Learn More
Waters Corp Waters Corporation SFC Column, Silica 2-EP, 19 x 100 mm, 5 μm
Catalog No. 50791251 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Viridis SFC columns are based on chromatographic silica production, designed for high reproducibility and predictability due to tight product specifications and very low metal content. They are suitable for analytical screening and preparative purification, allowing separation methods to be optimized and scaled up. The Viridis Silica 2-EP features 2-Ethylpyridine, widely used in achiral SFC separations for good retention, peak shape, and selectivity.
  • Designed for high reproducibility and predictability with tight product specifications and very low metal content
  • Suitable for analytical screening and preparative purification, allowing method optimization and scale-up
  • Features 2-Ethylpyridine chemistry for achiral SFC separations
  • Exhibits good retention, peak shape, and selectivity properties
  • Dimensions: OBD 19 x 100 mm
  • Particle size: 5 μm
Catalog No. 50-791-251 Supplier Waters Corp Supplier No. 186004944
Add to Cart
Add to Cart
These Viridis SFC columns are based on chromatographic silica production, designed for high reproducibility and predictability due to tight product specifications and very low metal content. They are suitable for analytical screening and preparative purification, allowing separation methods to be optimized and scaled up. The Viridis Silica 2-EP features 2-Ethylpyridine, widely used in achiral SFC separations for good retention, peak shape, and selectivity.
  • Designed for high reproducibility and predictability with tight product specifications and very low metal content
  • Suitable for analytical screening and preparative purification, allowing method optimization and scale-up
  • Features 2-Ethylpyridine chemistry for achiral SFC separations
  • Exhibits good retention, peak shape, and selectivity properties
  • Dimensions: OBD 19 x 100 mm
  • Particle size: 5 μm