Learn More
Waters Corp Waters SFC Column, Silica, OBD 30 x 75 mm, 5 μm, 100 Å
Supplier:  Waters Corp 186004921
Encompass_Preferred
This Viridis 5 μm Preparative SFC Column is a silica-based column designed for purification and separation methods. It features dimensions of OBD 30 × 75 mm.
  • Designed to be highly reproducible and predictable.
  • Tight product specifications.
  • Very low metal content.
  • Suitable for analytical screening and preparative column dimensions for purification.
  • Separation methods can be optimized and scaled up.
Catalog No. 50-791-235
Add to Cart
Request a Quote