Learn More
Waters Corp WATERS CORPORATION Viridis SFC Silica OBD Column, 30 x 50 mm, 5 μm
Catalog No. 50791234 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Viridis SFC Silica OBD Column, measuring 30 x 50 mm with a 5 μm particle size, offers new levels of reproducibility for laboratory purification. Suitable for both analytical and preparative chromatography, these columns ensure reliability and consistency, with chromatographic media setting the standard for quality.
  • Patented OBD™ technology ensures increased lifetime and reproducibility.
  • Optimized packing processes and unique column designs.
  • Optimum bed density achieved via axial force at the column inlet.
  • Predictable, uniform density profiles.
  • Chemically inert seals prevent leaks.
  • Highly reproducible with tight product specifications and low metal content.
  • Offers added selectivity due to highly polar, unbonded surface.
Catalog No. 50-791-234 Supplier Waters Corp Supplier No. 186004920
This Viridis SFC Silica OBD Column, measuring 30 x 50 mm with a 5 μm particle size, offers new levels of reproducibility for laboratory purification. Suitable for both analytical and preparative chromatography, these columns ensure reliability and consistency, with chromatographic media setting the standard for quality.
  • Patented OBD™ technology ensures increased lifetime and reproducibility.
  • Optimized packing processes and unique column designs.
  • Optimum bed density achieved via axial force at the column inlet.
  • Predictable, uniform density profiles.
  • Chemically inert seals prevent leaks.
  • Highly reproducible with tight product specifications and low metal content.
  • Offers added selectivity due to highly polar, unbonded surface.