Learn More
Waters Corp Waters Viridis Silica OBD Prep Column, 5 μm, 30 mm x 150 mm
Catalog No. 50791237 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Viridis Silica columns are engineered for high reproducibility and predictability, achieved through stringent product specifications and minimal metal content. They are suitable for both analytical screening and preparative purification, allowing optimization and scale-up of separation methods to preparative OBD columns.
  • Spherical silica particles: 5 μm particle size, 0.9 cc/g pore volume, 100 Å pore size, and 340 m²/g surface area.
  • High reproducibility and predictability from tight specifications and low metal content.
  • Suitable for both analytical screening and preparative purification.
  • Methods are scalable to preparative OBD columns.
  • Good retention, peak shape, and selectivity in achiral SFC separations, with or without additives.
Catalog No. 50-791-237 Supplier Waters Corp Supplier No. 186004923
Add to Cart
Add to Cart
Request a Quote
Viridis Silica columns are engineered for high reproducibility and predictability, achieved through stringent product specifications and minimal metal content. They are suitable for both analytical screening and preparative purification, allowing optimization and scale-up of separation methods to preparative OBD columns.
  • Spherical silica particles: 5 μm particle size, 0.9 cc/g pore volume, 100 Å pore size, and 340 m²/g surface area.
  • High reproducibility and predictability from tight specifications and low metal content.
  • Suitable for both analytical screening and preparative purification.
  • Methods are scalable to preparative OBD columns.
  • Good retention, peak shape, and selectivity in achiral SFC separations, with or without additives.