Learn More
Waters Corp Waters Viridis Column, Silica 2-Ethylpyridine, 100Å, 5 μm, 4.6 mm × 100 mm
Catalog No. 502037688 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Supercritical Fluid Chromatography (SFC) columns offer superior reproducibility for laboratory-scale purification. Their 2-Ethylpyridine chemistry ensures effective separation for diverse achiral SFC applications.
  • Advanced media manufacturing and column technology.
  • Delivers high reproducibility for purification.
  • Available as 2-Ethylpyridine and Silica columns for analytical and preparative chromatography.
  • Larger preparative formats feature patented Optimum Bed Density (OBD) technology.
  • 2-Ethylpyridine chemistry provides effective retention, peak shape, and selectivity for achiral SFC.
  • Silica columns offer high reproducibility and predictability due to tight specifications and low metal content.
Catalog No. 50-203-7688 Supplier Waters Corp Supplier No. 186004936
Add to Cart
Add to Cart
These Supercritical Fluid Chromatography (SFC) columns offer superior reproducibility for laboratory-scale purification. Their 2-Ethylpyridine chemistry ensures effective separation for diverse achiral SFC applications.
  • Advanced media manufacturing and column technology.
  • Delivers high reproducibility for purification.
  • Available as 2-Ethylpyridine and Silica columns for analytical and preparative chromatography.
  • Larger preparative formats feature patented Optimum Bed Density (OBD) technology.
  • 2-Ethylpyridine chemistry provides effective retention, peak shape, and selectivity for achiral SFC.
  • Silica columns offer high reproducibility and predictability due to tight specifications and low metal content.