Learn More
Waters Corp Waters Corporation Column, Viridis Silica 2-Ethylpyridine, 5 μm, 100 Å, 3 mm x 100 mm
Supplier:  Waters Corp 186005801
Encompass_Preferred
Designed and tested for SFC, the Viridis 5 μm columns offer a wide range of selectivity for increased resolution. They deliver excellent peak shapes under SFC conditions without mobile-phase additives, even for well-retained basic achiral compounds, due to the reduction and control of surface silanol activity on Viridis Hybrid particles.
  • Designed and tested for SFC.
  • Demonstrates a wide range of selectivity for greater resolution.
  • Features reduction and control of surface silanol activity on hybrid particles.
  • Delivers excellent peak shapes under SFC conditions without the use of mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.
Catalog No. 50-466-273