Learn More
Waters Corp Waters SFC Column, Viridis Silica, 100Å, 5 μm, 3 mm x 250 mm
Catalog No. 50466279 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Viridis Silica SFC column is designed for achiral SFC separations. It provides excellent retention, peak shape, and selectivity. The column is built upon a strong foundation of chromatographic silica production, ensuring high reproducibility and predictability. It adheres to strict product specifications and contains very low metal content.
  • Widely used in achiral SFC separations
  • Exhibits good retention, peak shape, and selectivity properties
  • Based on extensive chromatographic silica production expertise
  • Designed for high reproducibility and predictability
  • Features tight product specifications
  • Contains very low metal content
  • Available for analytical screening
  • Suitable for preparative column dimensions for purification
  • Separation methods can be optimized and scaled up to Viridis Prep OBD™ Columns
Catalog No. 50-466-279 Supplier Waters Corp Supplier No. 186005807
Add to Cart
Add to Cart
This Viridis Silica SFC column is designed for achiral SFC separations. It provides excellent retention, peak shape, and selectivity. The column is built upon a strong foundation of chromatographic silica production, ensuring high reproducibility and predictability. It adheres to strict product specifications and contains very low metal content.
  • Widely used in achiral SFC separations
  • Exhibits good retention, peak shape, and selectivity properties
  • Based on extensive chromatographic silica production expertise
  • Designed for high reproducibility and predictability
  • Features tight product specifications
  • Contains very low metal content
  • Available for analytical screening
  • Suitable for preparative column dimensions for purification
  • Separation methods can be optimized and scaled up to Viridis Prep OBD™ Columns