Learn More
Waters Corp OBD Prep Column, Silica, 5 μm, 100 Å, 10 mm x 50 mm
Catalog No. 502391661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Supercritical Fluid Chromatography (SFC) columns combine state-of-the-art media manufacturing with industry-leading column technology to provide a new level of reproducibility for laboratory scale purification. These SFC columns are available for analytical and preparative chromatography in various forms. All semi-prep and preparative column formats utilize Optimum Bed Density (OBD) Technology to enhance mechanical stability and extend column lifetimes.
  • Combines state-of-the-art media manufacturing with industry-leading column technology
  • Brings a new level of reproducibility to laboratory scale purification
  • Available for analytical and preparative chromatography
  • Semi-prep and preparative column formats are manufactured using Optimum Bed Density (OBD) Technology
  • OBD Technology provides mechanical stability and extended column lifetimes
Catalog No. 50-239-1661 Supplier Waters Corp Supplier No. 186008228
Supercritical Fluid Chromatography (SFC) columns combine state-of-the-art media manufacturing with industry-leading column technology to provide a new level of reproducibility for laboratory scale purification. These SFC columns are available for analytical and preparative chromatography in various forms. All semi-prep and preparative column formats utilize Optimum Bed Density (OBD) Technology to enhance mechanical stability and extend column lifetimes.
  • Combines state-of-the-art media manufacturing with industry-leading column technology
  • Brings a new level of reproducibility to laboratory scale purification
  • Available for analytical and preparative chromatography
  • Semi-prep and preparative column formats are manufactured using Optimum Bed Density (OBD) Technology
  • OBD Technology provides mechanical stability and extended column lifetimes