Learn More
Waters Corp Preparative SFC Column, Viridis Silica, 5 μm, 10 × 250 mm
Catalog No. 502390649 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis Silica column with a 5 μm particle size is a Preparative SFC Column. It has dimensions of OBD 10 × 250 mm. Viridis Silica Columns are designed for achiral SFC separations and are known for exhibiting good retention, peak shape, and selectivity properties.
  • Highly reproducible and predictable due to tight product specifications.
  • Very low metal content.
  • Available for both analytical screening and in preparative dimensions for purification.
  • Allows for optimization and scaling up of separation methods to preparative OBD columns.
Catalog No. 50-239-0649 Supplier Waters Corp Supplier No. 186008231
Add to Cart
Add to Cart
The Viridis Silica column with a 5 μm particle size is a Preparative SFC Column. It has dimensions of OBD 10 × 250 mm. Viridis Silica Columns are designed for achiral SFC separations and are known for exhibiting good retention, peak shape, and selectivity properties.
  • Highly reproducible and predictable due to tight product specifications.
  • Very low metal content.
  • Available for both analytical screening and in preparative dimensions for purification.
  • Allows for optimization and scaling up of separation methods to preparative OBD columns.