Learn More
Cayman Chemical VIrIdItoxIn 500ug
SDP
Supplier:  Cayman Chemical 26601500 UG
Encompass_Preferred
A mycotoxin; active against MSSA and MRSA, tetracycline-sensitive and -resistant Staphylococcus, vancomycin-sensitive and -resistant Enterococcus, and penicillin-sensitive and -resistant S. pneumoniae (MICs = 2-32 μg/ml); active against fish pathogens, including S. iniae and S. parauberis (MICs = 0.16-0.21 μg/ml); inhibits polymerization and the GTPase activity of E. coli FtsZ (IC50s = 8.2 and 7 μg/ml, respectively); inhibits proliferation of human DU145, LNCaP, and PC3 prostate cancer cells (IC50s = 5.36, 0.63, and 7.6 μM, respectively)
Catalog No. 50-293-5234