Learn More
Syndel USA Syndel Virkon Aquatic 10lb Tub
SDP
Catalog No. NC0398532
Encompass
Virkon Aquatic is a concentrated disinfectant powder that when mixed with water forms a powerful cleaning and disinfecting solution with efficacy against fish viruses, bacteria, fungi, and molds. Virkon Aquatic is EPA registered (except in California where registration is pending) for the disinfection of environmental surfaces associated with aquaculture including vehicles (boats, trailers, autos, etc.), nets, boots, waders, dive suits, hoses, brushes, hard surfaces and other similar equipment. Virkon Aquatic solutions are fast acting, colorless, fragrance free and non-irritating, which makes it the ultimate disinfectant for use in Aquatic applications.
This item is not returnable. View return policy
Catalog No. NC0398532 Supplier Syndel USA Supplier No. VIRKDLB0010
Virkon Aquatic is a concentrated disinfectant powder that when mixed with water forms a powerful cleaning and disinfecting solution with efficacy against fish viruses, bacteria, fungi, and molds. Virkon Aquatic is EPA registered (except in California where registration is pending) for the disinfection of environmental surfaces associated with aquaculture including vehicles (boats, trailers, autos, etc.), nets, boots, waders, dive suits, hoses, brushes, hard surfaces and other similar equipment. Virkon Aquatic solutions are fast acting, colorless, fragrance free and non-irritating, which makes it the ultimate disinfectant for use in Aquatic applications.