Learn More
MWI VETERINARY VIRKON S 10LB PAIL
SDP
Catalog No. NC0460167
Encompass_Preferred
Balanced, Stabilized Blend Of Peroxygen Compounds, Surfactant, Organic Acids And Inorganic Buffer. Recommended For Use As A Hard Surface Disinfectant In Livestock Production And Transportation Facilities. Kills Gram-Negative And Gram-Positive Bacteria, Viruses And Fungi Application. Flooding, Immersion, Mopping Or Spraying
Catalog No. NC0460167 Supplier MWI VETERINARY Supplier No. 036248
Balanced, Stabilized Blend Of Peroxygen Compounds, Surfactant, Organic Acids And Inorganic Buffer. Recommended For Use As A Hard Surface Disinfectant In Livestock Production And Transportation Facilities. Kills Gram-Negative And Gram-Positive Bacteria, Viruses And Fungi Application. Flooding, Immersion, Mopping Or Spraying